missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phosphite, 98%, Thermo Scientific Chemicals

Product Code. 10625342 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10625342

Brand: Thermo Scientific Acros 114500010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 762-04-9
MDL Number MFCD00044573
InChI Key LXCYSACZTOKNNS-UHFFFAOYSA-N
Synonym diethyl phosphite, diethyl phosphonate, phosphonic acid, diethyl ester, phosphonic acid diethyl ester, diethoxyphosphine oxide, hydrogen diethyl phosphite, diethyl acid phosphite, o,o-diethyl phosphonate, phosphorous acid, diethyl ester, ethyl phosphonate eto 2hpo
PubChem CID 6327654
SMILES CCO[P+](=O)OCC

Specifications

Color Colorless
Density 1.0800g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C4H10O3P+
Linear Formula (C2H5O)2P(O)H
Refractive Index 1.4070 to 1.4090 (20°C, 589nm)
Quantity 1 kg
Beilstein 01, 330
Fieser 01,251; 02,132; 12,187
IUPAC Name diethoxy(oxo)phosphanium
Molecular Weight (g/mol) 137.095
Formula Weight 138.1
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Diethyl phosphite, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl phosphite
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.