missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Marine Broth
Click to view available options
Quantity:
10 kg
Unit Size:
10kg
Specifications
Specifications
| Broth Type | Marine Broth |
| Color | Light beige |
| Form | Solid |
| Media Type | Dehydrated |
| Product Type | Microbiology Media |
| For Use With (Application) | For cultivating heterotrophic marine bacteria |
| Quantity | 10 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction