Learn More
BD Difco™ Select APS LB Broth Base
Brand: BD 292438
Additional Details : Weight : 1.00000kg
Specifications
Select APS LB Broth Base | |
Powder | |
Microbiology Media | |
500 g |
Beige | |
Dehydrated | |
Propagation and maintenance of E. coli for molecular biology. |