Learn More
Masterflex™ Differential Pressure Flowmeters
Product Code. 15202986
Change view
Click to view available options
Flow Rate Range:
0 to 10 LPM
0.05 to 5 LPM
2 to 200 mL/min.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Range unitSize
15202986 0 to 10 LPM Each
15305184 0.05 to 5 LPM Each
12058588 2 to 200 mL/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15202986 Supplier Masterflex™ Supplier No. 3290849
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Regulatory: This product is under eco-contribution tax.
Specifications
Type Differential Pressure Flowmeter
Accuracy ±2% (Full Scale)
Float Material Black Glass
Pressure Rating 100 psig
Connections 1/8 in. NPT (F)
Flow Rate Range 0 to 10 LPM
For Use With (Application) Achieve accurate, ultrafast volumetric measurement and control
Repeatability ±2%
Temperature (Metric) 50°C