Learn More
Medline Scientific™ Digital Camera
Product Code. p-8009740
Click to view available options
Resolution:
2048 x 1536
Unit Size:
Each
Product Code. 11980299
Brand: Medline Scientific™ 9556.3000M
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
For Use With (Equipment) microscopy
Pixels 3 MP
Port Type USB 2.0
Power Supply Details USB powered, 2.5w
Quantity 1 Ea.
Resolution 2048 x 1536
Sensor Types 0.5in Sensor