Learn More
TFA Dostmann™ Digital Indoor/Outdoor Thermo-Hygrometer
Product Code. 11808262
Change view
Click to view available options
Temperature Range (Metric):
-10°C to +60°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) unitSize
11808262 -10°C to +60°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11808262 Supplier TFA Dostmann™ Supplier No. 30.5013
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
For the simultaneous monitoring of indoor and outdoor climates.
  • Monitoring of both indoor and outdoor temperature and humidity
  • Measurement of outdoor values via cable sensor
  • Max/Min values
  • Wall-mounting or free-standing
Specifications
Type Thermohygrometer
Humidity Range 10 to 99%
Temperature Range (Metric) -10°C to +60°C
Battery Type 1 x AAA 1.5 V
Display Type Digital
Sensors Cable Sensor
Dimensions (L x W x H) 102 x 21 x 110 mm
Weight (Metric) 144 g
Voltage 1.5 V
For Use With (Application) For inside and outside measurement
Includes Thermo-hygrometer, 1 x 1.5 V AAA battery, instruction manual
Certifications/Compliance CE, EK-EL