Learn More
Ohaus™ Digital Multi-Tube Vortex Mixer VXMTDG
Product Code. 15797217 Shop All Ohaus Products
Click to view available options
Orbit:
3.6 mm
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 15797217
Brand: Ohaus™ 30392173
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
OHAUS™ Digital Multi-Tube Vortex Mixer VXMTDG is built sturdy to provide a stable and reliable vortexing action.
  • Ideal for applications requiring accuracy and repeatability
  • Process up to 50 samples at a time
  • LED displays for speed and time
  • Pulsing mode
TRUSTED_SUSTAINABILITY
Specifications
Orbit 3.6 mm
Load Bearing Capacity (Metric) 4.5 kg
Speed Range 500 to 2500 rpm
Electrical Requirements 230 V, 50/60 Hz, 1 A
Amperage 1 A
Dimensions (L x W) 24.10 x 38.40 cm
Height (English) 16.00 in.
Height (Metric) 40.6 cm
Length (English) 9.50 in.
Length (Metric) 24.10 cm
Width (Metric) 38.40 cm
Description Vortex Mixer, Multi-Tube, VXMTDG, EU
Model VXMTDG