Learn More
TFA Dostmann Digital Professional Thermometer with Cable Probe LT 202
The Digital professional thermometer with cable probe LT 202 is a professional tool. Its display shows the current temperature, the maximum and minimum values. For flexible placement the long waterproof cable sensor is perfect. This thermometer also has a temperature alarm.
Brand: TFA Dostmann 30.1052.02
Additional Details : Weight : 0.07900kg
Description
This handy measuring device is a professional tool for anyone for whom the correct temperature in refrigerators and freezers, aquariums or similar areas is a top priority. This digital professional thermometer works with high-quality sensor technology to display the temperature with great accuracy in a measuring range of -40°C to 70°C and to sound an alarm within seconds. This happens when the temperature exceeds or falls below the set alarm limits. This professional thermometer then emits a visual and acoustic signal. You can set the volume of this to one of two levels or mute it.
It is intuitive and simple to operate: Its clear display constantly shows the current temperature as well as the maximum and minimum values. These can be reset at the touch of a button. Thanks to its approx. two-metre-long waterproof cable sensor, you can also flexibly position the temperature sensor in damp environments or in water. The display unit can be attached magnetically to the outside of your freezer or refrigerator or fixed to the wall with a nail using the hanging loop. If you want to set up this professional thermometer at your workplace, it is best to use its integrated stand.
- Waterproof cable sensor with approx. 200 cm cable
- Can be used for a wide range of applications
- High accuracy (± 0,3° from -10°C to +20°C, ± 0,5° from -20°C to -10°C and +20°C to +40°C, ± 1° from -40°C to -20°C, +40°C to +70°C)
- Visual and acoustic temperature alarm (3 volume levels - loud, normal, mute)
- According to EN13485, ideal for professional use
- Permanent indication of max.-min. values
- With magnet, table stand and hanging slot
Specifications
Plastic | |
Yes | |
LCD |