Learn More
Aalborg™ Digital Thermal Mass Flow Controller
Click to view available options
Type:
Digital Regulator
Unit Size:
Each
Description
- Digital and analog modes operate
- Programmable flow configurations
- Available flow ranges starting from 0 to 10 sccm up to 0 to 100 slpm
- Maximum pressure of 1000 psig (70 bars)
- Multi-drop capability of up to 256 units
- Stores calibration data for up to 10 gases
- Totalizer indicates total gas quantity
- Alarm limits for high and low gas flow
- Conversion factors for up to 256 gases
- Auto tune function for optimum control response
- Self diagnostic test
Specifications
Specifications
| Type | Digital Regulator |
| Airflow Rate | 5 LPM |
| Pressure Rating | 70 bar |
| Flow Rate Range | 0 to 5 LPM |