missing translation for 'onlineSavingsMsg'
Learn More

Dihydro-ergotoxine Methanesulfonate, TRC

Product Code. 30135747
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135747 500 mg 500mg
30135090 250 mg 250mg
30006976 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135747 Supplier Toronto Research Chemicals Supplier No. TRCD450015500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8067-24-1
Formula Weight 2713.2268
InChI Formula InChI=1S/C31H41N5O5.CH4O3S/c1-16(2)26-28(38)35-11-7-10-24(35)31(40)36(26)29(39)30(41-31,17(3)4)33-27(37)19-12-21-20-8-6-9-22-25(20)18(14-32-22)13-23(21)34(5)15-19;1-5(2,3)4/h6,8-9,14,16-17,19,21,23-24,26,32,40H,7,10-13,15H2,1-5H3,(H,33,37);1H3,(H,2,3,4)/t19-,21?,23-,24-,26-,30+,31-;/m1./s1
IUPAC Name (6aR,9R)-N-((2R,5S,10aS,10bS)-10b-hydroxy-2,5-diisopropyl-3,6-dioxooctahydro-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl)-7-methyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinoline-9-carboxamide methanesulfonic acid
Molecular Formula C32H45N5O8S(R=CH(CH3)2), C36H45N5O8S(R=CH2Ph), C 34H47N5O8S (R=CH2CH(CH3)2, or CH(CH3)CH2CH3)
Molecular Weight (g/mol) 2053.46
Synonym Dihydro-ergotoxine Monomethanesulfonate, Hydergine, 8H-Oxazolo[3,2-a]pyrrolo[2,1-c]pyrazine, ergotoxine deriv., Indolo[4,3-fg]quinoline, ergotoxine deriv., CCK 179, Circanol, Co-dercrine mesylate, Co-dergocrine mesylate, Codergine mesylate, Codergocrine methanesulfonate, Coristin, Dacoren, Deapril ST, Decril, Dihydroergotoxin mesylate, Dihydroergotoxin methanesulfonate, Dihydroergotoxine mesylate, Dihydroergotoxine methanesulfonate, Dulcion, Ergodesit, Ergohydrin, Ergoloid mesylates, Ergoplus, Hydergin, Hydergyne, Ischelium, Novofluen, Orphol, Perenan, Progeril, Redergin, Sponsin, Trigot
Chemical Name or Material Codergocrine Mesylate

Specifications

Quantity 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.