missing translation for 'onlineSavingsMsg'
Learn More

Diiodobis(triphenylphosphine)nickel(II), Thermo Scientific Chemicals

Catalog Number p-7049709
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
82308-17-6
C36H30I2NiP2
837.09
MFCD00015866
CRLSSYRKSIEFOQ-UHFFFAOYSA-L
diiododbis triphenylphosphinenickel ii, diiodobis triphenylphosphine nickel ii, nii2 pph3 2, diiodonickel; triphenylphosphane
6102125
diiodonickel;triphenylphosphane
I[Ni++]I.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
82308-17-6
C36H30I2NiP2
837.09
MFCD00015866
CRLSSYRKSIEFOQ-UHFFFAOYSA-L
diiododbis triphenylphosphinenickel ii, diiodobis triphenylphosphine nickel ii, nii2 pph3 2, diiodonickel; triphenylphosphane
6102125
diiodonickel;triphenylphosphane
I[Ni++]I.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Linear Formula [(C6H5)3P]2NiI2
Quantity 2 g
Sensitivity Air and light sensitive
Formula Weight 837.08
Physical Form Crystalline Powder
Chemical Name or Material Diiodobis(triphenylphosphine)nickel(II)
compliance-icons compliance-icons
Product Identifier
  • Diiodobis(triphenylphosphine)nickel(II)
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Carcinogenicity Category 1B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'