missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl methylphosphonate, 90%, Thermo Scientific Chemicals

Product Code. 13428658 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 13428658

Brand: Thermo Scientific Acros 449380050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1445-75-6
Molecular Formula C7H17O3P
Molecular Weight (g/mol) 180.18
MDL Number MFCD00015022
InChI Key WOAFDHWYKSOANX-UHFFFAOYSA-N
Synonym diisopropyl methylphosphonate, diisopropyl methanephosphonate, bis 1-methylethyl methylphosphonate, unii-56v3og5dc7, methylphosphonic acid diisopropyl ester, phosphonic acid, methyl-, bis 1-methylethyl ester, phosphonic acid, methyl-, diisopropyl ester, methylphosphonic acid bis 1-methylethyl ester, dsstox_cid_4051, dsstox_rid_77269
PubChem CID 3073
ChEBI CHEBI:77325
IUPAC Name 2-[methyl(propan-2-yloxy)phosphoryl]oxypropane
SMILES CC(C)OP(C)(=O)OC(C)C

Specifications

Density 0.9850g/mL
Boiling Point 50.0°C to 51.0°C (1.0 mmHg)
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)2C(H)O]2P(O)CH3
Quantity 5 g
Specific Gravity 0.985
Formula Weight 180.18
Percent Purity 90%
Chemical Name or Material Diisopropyl methylphosphonate
compliance-icons
Product Identifier
  • Diisopropyl methylphosphonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.