Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for V6 Pumps
Click to view available options
Volume (Metric):
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
5 mL
10 mL
25 mL
Unit Size:
Each
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Diluter/Dispenser Syringe for V6 Pumps
Specifications
Specifications
| Description | 60mm stroke, 1/4-28 UNF thread connection, 4.8mm hole in plunger |
| Volume (Metric) | 25 μL |
| For Use With (Equipment) | V6 Pump |