Learn More
Lenz Laborglasintrumente Dimroth Condensers for Extractors with GL Threads
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05246018
Additional Details : Weight : 5.00000kg
Description
- Dimroth condensers for extractors
- With GL threads Dimroth condensers for extractors
- For condensing vapours
- With NS cone
- Water connection: GL 14 thread (with two threaded plastic olives)
- Made of DURAN™ tubing
Specifications
NS 71/51 | |
Duran Glass |