Learn More
Thermo Scientific™ Dionex™ Specially Formulated PTFE Tubing
Product Code. 11362143 Shop All Thermo Scientific Products
Click to view available options
Description:
250 and 350 um ID
250 um ID; Low pressure
350 um ID; Low pressure
For Use With (Equipment):
Dionex UltiMate 3000 RSLC Nano System
Fused Silica Capillary Tubing
Unit Size:
Pack of 2
Pack of 5
Product Code. 11362143
Brand: Thermo Scientific™ 161030
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Thermo Scientific™ Dionex Specially Formulated PTFE Tubing is used as connector for fused silica tubing (nano and cap LC). Choose fitting dimensions for OD of fused silica tubing used.
Thermo Scientific™ Dionex Specially Formulated PTFE Tubing is used as connector for fused silica tubing (nano and cap LC).
TRUSTED_SUSTAINABILITY
Specifications
Description 350 um ID; Low pressure
For Use With (Equipment) Fused Silica Capillary Tubing
Type Capillary Tubing
Product Line nanoLC