Learn More
Thermo Scientific™ Dionex™ Zero Dead Volume Fitting
Shop All Thermo Scientific Products
Click to view available options
Diameter (English) Outer:
1/16 in.
Unit Size:
Each
Description
Thermo Scientific™ Dionex Zero Dead Volume Fittings are for older generation nano and capillary LC systems.
Specifications
Specifications
| Description | Zero dead volume fitting |
| For Use With (Application) | Chromatography |
| Type | Fittings |
| Diameter (English) Outer | 1/16 in. |
| Product Line | nanoLC |