missing translation for 'onlineSavingsMsg'
Learn More

Diphenylmethane, 99+%

Product Code. p-7025646
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11444787

Brand: Thermo Scientific Alfa Aesar B21256.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 101-81-5 | C13H12 | 168.24 g/mol

Diphenylmethane, is used as an adhesive chemical composition for making flexible laminates for use in food packaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylmethane, is used as an adhesive chemical composition for making flexible laminates for use in food packaging.

Solubility
Soluble in ethanol, ether, benzene and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-81-5
Molecular Formula C13H12
Molecular Weight (g/mol) 168.24
MDL Number MFCD00004781
InChI Key CZZYITDELCSZES-UHFFFAOYSA-N
Synonym diphenylmethane, ditan, ditane, benzene, 1,1'-methylenebis, diphenyl methane, benzene, benzyl, methane, diphenyl, 1,1'-dimethylenebis benzene, benzene, phenylmethyl, methylenedibenzene
PubChem CID 7580
ChEBI CHEBI:38884
IUPAC Name benzylbenzene
SMILES C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 22°C to 26°C
Density 1.01
Boiling Point 262°C to 264°C
Flash Point 130°C (266°F)
Refractive Index 1.577
Quantity 100 g
UN Number UN3077
Beilstein 1904982
Merck Index 14,3328
Solubility Information Soluble in ethanol,ether,benzene and chloroform.
Formula Weight 168.24
Percent Purity ≥99%
Chemical Name or Material Diphenylmethane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.