missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford DIPSO
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Substance Name: 3-(N,N-Bis(2-Hydroxyethyl)amino)-2-hydroxypropane sulfonic acid
CAS Number: 68399-80-4
Molecular Formula: C7H17NO6S
Molecular Weight: 243.3
Chemical Purity: >98%
Soluble in:Water
Store at: Room TemperatureSpecifications
Specifications
| Chemical Name or Material | DIPSO |
| CAS | 68399-80-4 |
| Quantity | 100 g |
| Molecular Formula | C7H17NO6S |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction