Learn More
Lovibond™ Comparator Disc
Click to view available options
Analytes:
Ammonia
Chlorine
Chlorine (DPD)
Dissolved Oxygen
Hydrazine
Iron
Nitrate
Ortho-phosphate
ph Value
Description:
Disc 2/1B
Disc 2/1E
Disc 2/1H
Disc 2/1J
Disc 3/113
Disc 3/116
Disc 3/142
Disc 3/2 Hypo
Disc 3/40A
Disc 3/40B
Disc 3/40K
Disc 3/70
Disc NAB
Disc NOE
Disc: NOH
missing translation for 'unitSize'
Each
Pack of 2000
Description
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
- Easy to use, visual assessment of the sample; simply rotate disc to match color
- Glass, fade-free standards for longevity of use
- Shock resistant, scratch resistant
- Optional lighting units for guaranteed lighting conditions 24 hours/day
- Optional Nessleriser systems for longer path lengths
- More than 400 color discs are available for the Comparator 2000+
- Lovibond™ color standards consist exclusively of through-and-through colored and flat-ground glasses
Specifications
Specifications
| Analytes | Iron |
| Description | Disc 3/116 |
| For Use With (Equipment) | Comparator 2000+ |
| Range | 0.1, 0.2, 0.3, 0.4, 0.5, 0.6, 0.7, 0.8, 1.0mg/L |
| Type | Color Comparator Disc |