Learn More
Kartell™ Discharge Tube for Dosipet
Click to view available options
For Use With (Equipment):
Dosipet 2.5 to 10mL
Dosipet 25 to 100mL
Unit Size:
Each
Description
25-100mL for Dosipet dispenser
Specifications
Specifications
| Description | Discharge tube |
| Material | FEP |
| For Use With (Equipment) | Dosipet 25 to 100mL |