Learn More
Kartell™ Discharge Tube for Dosipet
Product Code. 13111104
Click to view available options
For Use With (Equipment):
Dosipet 2.5 to 10mL
Dosipet 25 to 100mL
Unit Size:
Each
Product Code. 13111104
Brand: Kartell™ 8301700
Only null left
Add to basket
Add to basket
Kartell™ Discharge Tube for Dosipet is an accessory for Dosipet products, which are used to measure out a variety of solutions and solvents.
2.5-10mL for Dosipet dispenser
Specifications
Description Discharge tube
Material FEP
For Use With (Equipment) Dosipet 2.5 to 10mL