Learn More
Agilent BioTek Dispense Bottle, 4L with Filter & Tubing
Dispense Bottle, 4L with Filter & Tubing is for use with Microplate Washers and Readers
Brand:  Agilent BioTek 4070076
Additional Details : Weight : 0.90000kg
Product Code. 16502192
  • 202.91€ / Each
Estimated Shipment: 02-08-2024
to see stock.
Specifications
Dispense Bottle
4 L
Agilent BioTek Microplate Washer and Reader Instruments