Learn More
BRAND™ Bottletop Dispenser Thread Adapters
Shop All Brand Products
Click to view available options
Material:
ETFE
PTFE
Polypropylene
Stainless Steel
Thread Size:
2 in. (Outer), 3/4 in. (Inner)
32 GL (inner thread)
32 GL (outer thread)
GL 32/38
GL 45/35
GL 45/38
Unit Size:
Each
Description
- Constructed of Stainless Steel, PTFE, ETFE, or polypropylene.
- For use with Dispensette™, seripettor™, Titrette™, or QuikSip™ bottletop dispensers.
- Available in a variety of thread sizes.
Specifications
Specifications
| Product Type | Dispenser Thread Adapter |
| Material | PTFE |
| For Use With (Application) | Dispensette Direct Mounting on Drum |
| For Use With (Equipment) | DispensetteIII, Dispensette Organic Remote Dispensing Unit |
| Quantity | 1 Ea. |
| Thread Size | 32 GL (inner thread) |