Learn More
Kinematica Dispersing Vessel
Product Code. 15883744 Shop All Kinematica Products
Change view
Click to view available options
Capacity (Metric):
150 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15883744 150 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15883744 Supplier Kinematica Supplier No. 11051041
Only null left
Add to basket
Add to basket
Cloverleaf internal shape effectively interrupts the flow in the radial direction and generates additional counteracting force.
  • Cloverleaf internal shape
  • Interrupts flow in radial direction to generate additional counteracting forces
  • High turbulences maximize mixing and dispersion efficiency
  • Minimizes processing time and energy output
Specifications
Capacity (Metric) 150 mL
Size Thread: GL45
Bore Size (Metric) 20 mm
Diameter (Metric) 54 mm
Height (Metric) 150 mm
Model GS 25KL2