Learn More
Terracycle™ Disposable Gloves Zero Waste Box™
Recycle Disposable Gloves using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
Brand: Terracycle™ CS45-M-BE
Additional Details : Weight : 1.00000kg
Description
Use this Zero Waste Box to recycle:
- Vinyl, nitrile, and latex gloves
Approximate waste units accepted:
- Small: 600 gloves
- Medium: 1700 gloves
- Large: 3100 gloves
Do not send in:
- Medical waste
- This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, home care, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
- Autoclaved material
- Gloves that are contaminated with materials considered hazardous or dangerous
- Heavy duty, fabric, and leather gloves
- Hairnets, earplugs, and lab coats or garments
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) or material contaminated with blood which present a danger to the environment, or to people
- Batteries, pressurized canisters, broken glass
Specifications
15 kg | |
Beige, Green, Red, White | |
Single-use | |
Medium | |
Not Autoclavable | |
Recycling Box |
33 lb. | |
30.5 x 30.5 x 84 cm | |
Yes | |
Non-sterile | |
Cardboard, Plastic |