missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Disposable Healthcare Tunic and Pants Set, 48 g/m2

Product Code. 17730169 Shop All Medicom Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 60
This item is not returnable. View return policy

Product Code. 17730169

Brand: Kolmi Hopen™ J40480/L

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

These sets of disposable tunics and pants provide extra protection for healthcare workers.

  • Light and breathable polypropylene material
  • Loose shape allows user to maintain good movement
  • Collarless shirt
  • Pants with elasticated waist
  • Individual packaging for hygiene and controlled distribution
  • 48 g/m2 fabric weight

Specifications

Size Large
Material Polypropylene
Color Navy
Sleeve Style Short Sleeve
Disposable Single-use
For Use With (Application) Dental, Medical
No. per Pack 60 Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.