Learn More
Copan Italia™ Disposable Inoculation Loops
Click to view available options
Color:
Dark Blue
Light Blue
Quantity:
2000/Pk.
4000/Pk.
500/Pk.
Unit Size:
Pack of 2000
Pack of 4000
Pack of 500
Description
- Sterile
- Calibrated
- Rigid plastic
Specifications
Specifications
| Disposable | Single-use |
| Material | Rigid Plastic |
| Sterility | Sterile |
| Color | Light Blue |
| Product Type | Inoculating Loop |
| For Use With (Equipment) | Laboratory and Research use |
| Quantity | 2000/Pk. |
| For Use With (Application) | Laboratory and Research use |