Learn More
Simport™ Disposable Transfer Pipets
Click to view available options
Volume (Metric):
5 mL
6 mL
Unit Size:
Pack of 4000
Pack of 500
Description
- LDPE transfer pipets
- Put an end to matching rubber bulbs with glass pipets
- Can be sealed and refrigerated
- Work well whenever there is a need for quick, safe transfer of fluids
- Temperature resistant down to -196 °C. Can be gaz (EtO) sterilized
- Choose between 12 popular models, available in several sizes, tip designs and lengths, in sterile or non-sterile packaging
- Available with graduations
Specifications
Specifications
| Bulb Draw (Metric) | 2.3 mL |
| Diameter (Metric) Outer | 5 mm |
| Material | LDPE |
| Drops/mL | 25 |
| For Use With (Application) | For quick, safe transfer of fluids |
| Graduated | No |
| Quantity | 500 Pack |
| Volume (Metric) | 6 mL |
| Length (Metric) | 22.5 cm |
| Type | Transfer Pipet |
| Show More |