missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Photometer Tube Test

Product Code. 11953757
Click to view available options
Range:
0 to 15 mg/L
0 to 2 mg/L
0 to 20 mg/L
0 to 7 mg/L Zn, 0 to 35 mg/L Zn
0.2 to 20 mg/L
0.3 to 20 mg/L
No. of Tests:
25
30
Unit Size:
0.20kg
Pack of 25
Pack of 30
This item is not returnable. View return policy

Product Code. 11953757

Brand: Palintest™ PL 420

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Predispensed reagent tests require minimal handling and speed up processing. Palintest™; Photometer Tube Test reagents are supplied in 16mm tubes for improved resolution, with greater sensitivity in the lower part of the measurement range.

  • For ultimate convenience, the Palintest™ range of Tubetest reagents offer pre-dispensed reagents for minimal handling with barcoded reagent labelling for ease of use in wastewater applications.
  • For a complete wastewater analysis system, use Palintest™ tubetests with any Palintest™ multiparameter photometer and heater block.
  • Tubetest reagents are supplied in 16mm tubes for improved resolution, with greater sensitivity in the lower part of the measurement range.

Specifications

Analytes Ammonia
For Use With (Equipment) Photometer, heater block
No. of Tests 25
Quantity 24
Range 0 to 15 mg/L
Type Photometer tube test reagent
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • TEST TUBE AMMONIA 0 - 15 MG/L
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash contaminated skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P314-Get medical advice/attention if you feel unwell.
  • P321-Specific treatment (see medical advice on this label).
  • P363-Wash contaminated clothing before reuse.
  • P391-Collect spillage.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide, potassium iodide, mercury dichloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.