Learn More
Lenz Laborglasinstrumente™ Distilling Links
Distilling Links for the connection of distillation thermometers of 55 mm built-in length.
Brand: Lenz Laborglasinstrumente™ 05043004
Additional Details : Weight : 0.80000kg
Description
- With Liebig condenser
- Outlet with NS cone
- Water connection with glass olives, with NS socket
Specifications
Distilling Link | |
Duran Glass | |
NS 29/32 | |
350 mm |