missing translation for 'onlineSavingsMsg'
Learn More

Mesitylacetonitrile, 98%

Product Code. 11428907
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11428907

Brand: Thermo Scientific Alfa Aesar B20534.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 34688-71-6 | C11H13N | 159.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34688-71-6
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.232
MDL Number MFCD00013815
InChI Key SDKQOGSGNPGPRN-UHFFFAOYSA-N
Synonym mesitylacetonitrile, 2,4,6-trimethylphenylacetonitrile, 2-mesitylacetonitrile, 2-2,4,6-trimethylphenyl acetonitrile, 2,4,6-trimethylbenzyl cyanide, benzeneacetonitrile, 2,4,6-trimethyl, 2,4,6-trimethylphenyl acetonitrile, 2-2,4,6-trimethylphenyl ethanenitrile, mesitylacetonitrile #
PubChem CID 520698
IUPAC Name 2-(2,4,6-trimethylphenyl)acetonitrile
SMILES CC1=CC(=C(C(=C1)C)CC#N)C

Specifications

Melting Point 77°C to 80°C
Boiling Point 120°C (1 mmHg)
Quantity 25 g
UN Number UN3439
Beilstein 1942677
Formula Weight 159.23
Percent Purity 98%
Chemical Name or Material Mesitylacetonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.