missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-α-Phenylglycine, 99%

Catalog Number p-3758672
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
2835-06-5
C8H9NO2
151.16
MFCD00064402
ZGUNAGUHMKGQNY-UHFFFAOYNA-N
DL-α-Aminophenylacetic acid
2-amino-2-phenylacetic acid
NC(C(O)=O)C1=CC=CC=C1
This item is not returnable. View return policy
2835-06-5
C8H9NO2
151.16
MFCD00064402
ZGUNAGUHMKGQNY-UHFFFAOYNA-N
DL-α-Aminophenylacetic acid
2-amino-2-phenylacetic acid
NC(C(O)=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 290°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C6H5CH(NH2)CO2H
Quantity 100 g
Beilstein 14, 460
Merck Index 15, 7402
Specific Rotation -1° to +1° (20°C, 589nm) (c=1, 1N HCl)
Formula Weight 151.16
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material DL-α-Phenylglycine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'