missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Pipecolic Acid, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | DL-Pipecolic Acid |
| CAS | 535-75-1 |
| Molecular Formula | C6 H11 N O2 |
| Quantity | 10 g |
| Synonym | 2-Piperidinecarboxylic acid, Pipecolic acid (8CI), (RS)-2-Piperidinecarboxylic acid, (±)-2-Piperidinecarboxylic acid, (±)-Pipecolic acid, (±)-Pipecolinic acid, 2-Carboxypiperidine, 2-Pipecolinic acid, 2-Piperidinylcarboxylic acid, DL-2-Piperidinecarboxylic acid, DL-Pipecolic acid, DL-Pipecolinic acid, Dihydrobaikiane, Hexahydro-2-picolinic acid, Homoproline, NSC 17125, Pipecolinic acid, Piperidine-6-carboxylic acid, Piperolinic acid, α-Pipecolinic acid |
| InChI Formula | InChI=1S/C6H11NO2/c8-6(9)5-3-1-2-4-7-5/h5,7H,1-4H2,(H,8,9) |
| SMILES | OC(=O)C1CCCCN1 |
| IUPAC Name | piperidine-2-carboxylic acid |
| Molecular Weight (g/mol) | 129.16 |
| Formula Weight | 129.079 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction