missing translation for 'onlineSavingsMsg'
Learn More

DL-Serine, TRC

Product Code. 30415305
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
Product Code. Quantity unitSize
30415305 1 g -
30421091 50 g -
30416904 10 g 10g
3 options
This item is not returnable. View return policy

Product Code. 30415305

Brand: Toronto Research Chemicals TRCS2709801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-84-1
Formula Weight 105.0426
InChI Formula InChI=1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)
IUPAC Name 2-amino-3-hydroxypropanoic acid
Molecular Formula C3 H7 N O3
Molecular Weight (g/mol) 105.09
SMILES NC(CO)C(=O)O
Synonym Serine (9CI, ACI), DL-Serine (ZCI), Serine, DL- (8CI), (±)-Serine, 2-Azaniumyl-3-hydroxypropanoate, DL-Ser, NSC 9960
Chemical Name or Material DL-Serine

Specifications

Quantity 1 g
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.