missing translation for 'onlineSavingsMsg'
Learn More

DL-Tartaric acid, 99%, Thermo Scientific Chemicals

Catalog Number p-7080936
Click to view available options
Quantity:
250 g
500 g
1000 g
This item is not returnable. View return policy
133-37-9
C4H6O6
150.09
MFCD00071626
FEWJPZIEWOKRBE-UHFFFAOYNA-N
dl-tartaric acid, 2,3-dihydroxysuccinic acid, tartaric acid, racemic acid, uvic acid, traubensaure, paratartaric acid, paratartaric aicd, dl-tartrate, racemic tartaric acid
875
CHEBI:15674
2,3-dihydroxybutanedioic acid
OC(C(O)C(O)=O)C(O)=O
This item is not returnable. View return policy
133-37-9
C4H6O6
150.09
MFCD00071626
FEWJPZIEWOKRBE-UHFFFAOYNA-N
dl-tartaric acid, 2,3-dihydroxysuccinic acid, tartaric acid, racemic acid, uvic acid, traubensaure, paratartaric acid, paratartaric aicd, dl-tartrate, racemic tartaric acid
875
CHEBI:15674
2,3-dihydroxybutanedioic acid
OC(C(O)C(O)=O)C(O)=O

DL-Tartaric acid is used as a synergist for antioxidants, emulsifier, sequestrant and flavoring agent. It is also added with citric acid to prepare effervescent salts, thereby enhancing the taste of oral medications. It is also utilized in pigments, processing aids, ink, toner and colorant products. It acts as a chelating agent in metal and farming industries. Further, it is used as lubricant and grease. It is mixed with sodium bicarbonate and used as a leavening agent in food preparation. In the pharmaceutical industry, it is utilized in the preparation of tartar emetic, which is used in cough syrup as an expectorant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Tartaric acid is used as a synergist for antioxidants, emulsifier, sequestrant and flavoring agent. It is also added with citric acid to prepare effervescent salts, thereby enhancing the taste of oral medications. It is also utilized in pigments, processing aids, ink, toner and colorant products. It acts as a chelating agent in metal and farming industries. Further, it is used as lubricant and grease. It is mixed with sodium bicarbonate and used as a leavening agent in food preparation. In the pharmaceutical industry, it is utilized in the preparation of tartar emetic, which is used in cough syrup as an expectorant.

Solubility
Soluble in water, alcohol, mineral acids and alkalies.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY
Melting Point 210°C to 212°C (decomposition)
Beilstein 1725148
Merck Index 14,9069
Solubility Information Soluble in water,alcohol,mineral acids and alkalies.
Formula Weight 150.09
Percent Purity 99%
Quantity 250 g
Chemical Name or Material DL-Tartaric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'