missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DMF-dG Phosphoramidite, standard grade, 20-400 finish bottle

Product Code. 17967102 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
17967102 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 17967102

Brand: Thermo Scientific™ 27173710

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DMF-dG CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyguanosine (dG) with Dimethylaminoformamidine (DMF) base protection, for use in oligonucleotide synthesis.

DMF-dG CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyguanosine (dG) with Dimethylaminoformamidine (DMF) base protection, for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. By using Ac-dC, Bz-dA, and DMF-dG monomers, fast cleavage and deprotection can be effected by a 1:1 mixture of aqueous ammonium hydroxide and aqueous methylamine (or AMA) in 10 min. This product is supplied in a 30 mL Amber bottle with a 20-400 Finish neck style, which is compatible with most Expedite Systems of oligonucleotide synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N2-(dimethylaminoformamidine)-2'-deoxyguanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C43H53N8O7P
Base: dG
Base Protection: DMF
Formula weight: 824.9 g/mol
CAS number: 330628-04-1
HPLC Purity: ≥98.0%; M-11 Impurity ≤0.01%
31P NMR Purity: ≥98.0%; Sum of non-primary peaks at 140-152 ppm (PIII impurities): ≤1.5%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Expedite Synthesizers
Product Type DMF-dG CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Deoxyguanosine (dG)
Reagent Features Fast deprotection
Shipping Condition Room Temperature
Purity 98%
Quantity 1 g
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type DNA
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.