Learn More
Thermo Scientific™ DNA Marker, low range, 250 Base Pair Ladder
Product Code. 15416119
Click to view available options
Quantity:
1 mL
2 mL
Unit Size:
1mL
2mL
Product Code. 15416119
Brand: Thermo Scientific™ J63973.S0
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Molecular Weight Marker
Form Liquid
Quantity 2 mL