missing translation for 'onlineSavingsMsg'
Learn More

DNAI1 Antibody Pair, Novus Biologicals™

Product Code. 18377689 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377689 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377689 Brand Novus Biologicals Brand No. H00027019AP22

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody Pair

DNAI1 Polyclonal antibody specifically detects DNAI1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAI1
Applications Sandwich ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Sandwich ELISA 1 to 100 ng/mL
Formulation 1x PBS, pH 7.4
Gene Alias Axonemal dynein intermediate chain 1, CILD1MGC26204, dynein intermediate chain DNAI1, dynein, axonemal, intermediate chain 1, dynein, axonemal, intermediate polypeptide 1, ICS, ICS1, immotile cilia syndrome 1, PCDdynein intermediate chain 1, axonemal
Quantity 1 Set
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27019
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.