Learn More
Dobutamine hydrochloride, Thermo Scientific™
93.30€ - 253.65€
Chemical Identifiers
| CAS | 49745-95-1 |
|---|---|
| Molecular Formula | C18H24ClNO3 |
| Molecular Weight (g/mol) | 337.84 |
| MDL Number | MFCD00153795 |
| InChI Key | BQKADKWNRWCIJL-UHFFFAOYNA-N |
| Synonym | dobutamine hydrochloride, dobutamine hcl, dobutrex, inotrex, dl-dobutamine hydrochloride, dobutamina clorhidrato spanish, 4-2-4-4-hydroxyphenyl butan-2-ylamino ethyl benzene-1,2-diol hydrochloride, +--4-2-3-p-hydroxyphenyl-1-methylpropyl amino ethyl pyrocatechol hydrochloride, 1,2-benzenediol, 4-2-3-4-hydroxyphenyl-1-methylpropyl amino ethyl-, hydrochloride, +- |
| PubChem CID | 65324 |
| ChEBI | CHEBI:4671 |
| IUPAC Name | hydrogen 4-(2-{[4-(4-hydroxyphenyl)butan-2-yl]amino}ethyl)benzene-1,2-diol chloride |
| SMILES | [H+].[Cl-].CC(CCC1=CC=C(O)C=C1)NCCC1=CC=C(O)C(O)=C1 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15463939
|
Thermo Scientific Alfa Aesar
J61333.MD |
250mg |
93.30€
250mg |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15453939
|
Thermo Scientific Alfa Aesar
J61333.03 |
1g |
282.00€ 253.65€ / 1g Save 28.35€ 10% Off |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsDobutamine hydrochloride is a catecholamine that acts as a β-adrenergic receptor agonist with potent positive inotropic effects in vivo. It has its strongest effects on β1 receptors, with lesser but significant β2 receptor activation and only weak α1 receptor actions. As a result, it strongly increases cardiac contractility with modest chronotropic, arrhythmogenic, and vascular side effects. It is also a polyphenolic antioxidant and can inhibit all human carbonic anhydrase isoforms with Ki values near 1 μM.
Solubility
Soluble in water or ethanol with warming. Also soluble in methanol
Notes
Solutions oxidize readily at pH greater than 11. Protect solutions from light
Chemical Identifiers
| 49745-95-1 | |
| 337.84 | |
| BQKADKWNRWCIJL-UHFFFAOYNA-N | |
| 65324 | |
| hydrogen 4-(2-{[4-(4-hydroxyphenyl)butan-2-yl]amino}ethyl)benzene-1,2-diol chloride |
| C18H24ClNO3 | |
| MFCD00153795 | |
| dobutamine hydrochloride, dobutamine hcl, dobutrex, inotrex, dl-dobutamine hydrochloride, dobutamina clorhidrato spanish, 4-2-4-4-hydroxyphenyl butan-2-ylamino ethyl benzene-1,2-diol hydrochloride, +--4-2-3-p-hydroxyphenyl-1-methylpropyl amino ethyl pyrocatechol hydrochloride, 1,2-benzenediol, 4-2-3-4-hydroxyphenyl-1-methylpropyl amino ethyl-, hydrochloride, +- | |
| CHEBI:4671 | |
| [H+].[Cl-].CC(CCC1=CC=C(O)C=C1)NCCC1=CC=C(O)C(O)=C1 |
Specifications
| 49745-95-1 | |
| MFCD00153795 | |
| dobutamine hydrochloride, dobutamine hcl, dobutrex, inotrex, dl-dobutamine hydrochloride, dobutamina clorhidrato spanish, 4-2-4-4-hydroxyphenyl butan-2-ylamino ethyl benzene-1,2-diol hydrochloride, +--4-2-3-p-hydroxyphenyl-1-methylpropyl amino ethyl pyrocatechol hydrochloride, 1,2-benzenediol, 4-2-3-4-hydroxyphenyl-1-methylpropyl amino ethyl-, hydrochloride, +- | |
| BQKADKWNRWCIJL-UHFFFAOYNA-N | |
| hydrogen 4-(2-{[4-(4-hydroxyphenyl)butan-2-yl]amino}ethyl)benzene-1,2-diol chloride | |
| 65324 | |
| 337.85 |
| C18H24ClNO3 | |
| 14,3395 | |
| Soluble in water or ethanol with warming; Also soluble in methanol | |
| [H+].[Cl-].CC(CCC1=CC=C(O)C=C1)NCCC1=CC=C(O)C(O)=C1 | |
| 337.84 | |
| CHEBI:4671 | |
| Dobutamine hydrochloride |
Safety and Handling
GHS H Statement
H361-H302-H312-H332
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
H302+H312+H332-H361
missing translation for 'einecsNumber' : 256-464-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.