missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecylbenzene sulfonic acid, mixture of C10-C13 isomers
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -10.0°C |
| Color | Brown |
| Density | 1.0600g/mL |
| Boiling Point | 315.0°C |
| Flash Point | 210°C |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Linear Formula | CH3(CH2)xC6H4SO3Hx=9to12 |
| Refractive Index | 1.5035 to 1.5065 |
| Quantity | 500 g |
| Show More |
Safety and Handling
Product Identifier
- Dodecylbenzene sulfonic acid, mixture of C10-C13 isomers
- Danger
- Skin corrosion/irritation Category 1 C
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction