missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Doripenem
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 364622-82-2
- Molecular Formula: C15H24N4O6S2 H2O
- Molecular Weight: 438.52
Specifications
Specifications
| Chemical Name or Material | Doripenem |
| CAS | 364622-82-2 |
| Quantity | 50 mg |
| Molecular Formula | C15H24N4O6S2 - H2O |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction