missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Dorsomorphin dihydrochloride
GREENER_CHOICE

Product Code. 18055202
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18055202

Brand: Tocris Bioscience™ 3093/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent AMPK inhibitor

Dorsomorphin dihydrochloride is a potent inhibitor of AMP-activated protein kinase (AMPK) (Ki = 109 nM). Displays no significant activity on several structurally related kinases including ZAPK, SYK, PKCθ, PKA and JAK3. Inhibits AMPK activation induced by AICAR (Cat. No. 2840) and Metformin (Cat. No. 2864). Dorsomorphin dihydrochloride also inhibits bone morphogenetic protein (BMP) type I receptors (ALK2, ALK3 and ALK6), promotes cardiomyogenesis in mouse embryonic stem cells (ESCs) in vitro and promotes neural differentiation of hPSCs as part of a chemical cocktail. Shown to induce autophagy in cancer cell lines via a mechanism independent of AMPK inhibition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1219168-18-9
Molecular Formula C24H27Cl2N5O

Specifications

Quantity 50 mg
Formula Weight 472.41
Purity >98%
Molecular Weight (g/mol) 472.414
Chemical Name or Material Dorsomorphin dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.