Learn More
Sampling Systems™ Double Bagged - USP Class VI Pallet Knives
Shop All Sampling Systems Products
Click to view available options
Length (Metric) Blade:
200 mm
Unit Size:
Pack of 40
Description
- The SteriWare™ Ultra Pallet Knives are commonly used in the manufacture of cosmetics, food and pharmaceuticals.
- They are also popular for levelling off scoops etc.
- The pallet knives all have an ergonomic handle and are designed to flex.
- The pallet knives are designed to be used straight from the bag.
- Made from food grade, FDA and EC compliant HDPE.
- HDPE has good chemical resistance and can be easily recycled.
- The pallet knives are sterilised by gamma irradiation.
- USP CLASS VI - High quality medical grade polymer that has also been independently tested to USP 661.
- Cleanroom Manufactured - The pallet knives are entirely made in cleanroom conditions with final packing carried out in a class 10,000 cleanroom.
- Each pallet knife is 100% inspected before packing.
Specifications
Specifications
| Sterility | Sterile |
| Length (Metric) Blade | 200 mm |
| Material | HDPE |