Learn More
BRAND™ Double-Ended PTFE Magnetic Stirring Bars
Excellent centering
Brand:  BRAND™ 137935
197.92 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 1.86000kg
 View more versions of this product
Product Code. 10668881
  • 235.00€ / Pack of 10
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
Description
Description
  • Small contact surface
  • High turbulence
  • Works well even at low speeds
  • Specifications
    Specifications
    8 mm
    10
    54 mm
    Dumbbell