Learn More
Fisherbrand™ Dual Scale Plain Form Hydrometer
Product Code. 11727073
Encompass_Preferred
Click to view available options
Length (Metric):
380 mm
Unit Size:
Each
Product Code. 11727073
Brand: Fisherbrand™ UN7200
Only null left
Add to basket
Add to basket
A dual scale, universal plain form hydrometer for measuring specific gravity/relative density (g/cm3) of liquids lighter than or heavier than water and Baume in various ranges.
  • Range of 0.7000 to 2.00
  • Lead-free ballast and glass
  • Individually serialized
  • Temperature standardization of 15°C
  • Used in the chemistry, pharmacology, food and beverage, and starch industries
  • Store in a clean, dry, safe place, not subject to extreme temperatures
Order Info
dimLengthMetric
Specifications
Quantity 1Each
Type Double Scale Hydrometer
Increments 0.050°
Length (Metric) 380 mm
Density (Metric) 50 g/L
Content And Storage Cool, dry, safe
Baume Range -10 to 70° (Less Dense Liquid Other than Water), -0 to 70° (Liquid Denser than Water)
For Use With (Application) Measuring the density of liquids both lighter and heavier than water
Specific Gravity Range 0.7 to 2.0%