Learn More
Kartell™ Douche Can
Kartell™ Douche Can is a tool for irrigation and is provided with a falt back and suspension tag.
7.75€ - 9.45€
Specifications
Description | Douche can or irrigator |
---|---|
Material | Polypropylene |
Shape | Cylindrical |
Type | Douche Can |
Diameter (Metric) | 9.9 mm |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12621006
|
Kartell™
0059000 |
2000 mL |
9.45€
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10396251
|
Kartell™
0058900 |
1000 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
Kartell™ Douche Can is a tool for irrigation and is provided with a falt back and suspension tag.
- Provided with flat back
- Includes suspension tag
- Autoclavable
Specifications
Douche can or irrigator | |
Cylindrical | |
9.9 mm |
Polypropylene | |
Douche Can |