missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom™ 50WX2 50-100 (H), Thermo Scientific Chemicals

Catalog Number p-7046319
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
12612-37-2
MFCD00132722
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
12612-37-2
MFCD00132722

AmberChrom™ 50WX2 50-100 (H), is used as a catalyst for selective monotetrahydropyranylation of symmetrical diols and for selective monoesterification of dicarboxylic acids. It is also used to remove calcium from aqueous samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex™ 50WX2 50-100 (H) is used as a catalyst for selective monotetrahydropyranylation of symmetrical diols and for selective monoesterification of dicarboxylic acids. It is also used to remove calcium from aqueous samples.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 50 g
Solubility Information Insoluble in water.
Chemical Name or Material Dowex™ 50WX2 50-100 (H)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'