Learn More
Nickel Electro™ Drain Tap for Clifton Water Baths
Product Code. 13127266 Shop All Nickel-Electro Products
Click to view available options
For Use With (Equipment):
Clifton Range Water Baths
Unit Size:
Each
Product Code. 13127266
Brand: Nickel Electro™ BX0953
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Drain Tap for Clifton water baths. These fast draining outlets makes emptying 50% faster than emptying without a tap.
  • Water baths with serial numbers greater than 75100 should order this accessory Drain Tap BX0953 - bigger bore - same fitting instructions as BX0688 detailed in above images.
  • These fast draining outlets means it only takes 13 minutes to empty a 28 litre bath, instead of 28 minutes, 50% quicker than before.
  • Before fitting make sure bath is empty of liquid. Unscrew the stop plug with a 6mm Allen key whilst holding the nut with a 21mm spanner or nut holder supplied with tap.
  • Screw the Tap into position, recommend pinch tight.
  • Instructions for fitting in download area.
  • Features:
    1. Providing a gated on/off valve to empty fluids conveniently from the bath.
    2. Nozzle supplied with tap for laboratory tubing.
Specifications
Description Drain tap
For Use With (Equipment) Clifton Range Water Baths
Material Stainless Steel