Learn More
Nickel Electro™ Draining Syphon
Shop All Nickel-Electro Products
Click to view available options
For Use With (Equipment):
NE4-8 D/CT, NE1B-28, NE1-2.5 E, NE4-28 D, NE4-22 T Stirred Digital Clear Water Baths
Unit Size:
Each
Description
- Water temperature must be below 45°C before use
- Allows the easy and quick emptying of water baths, immerse into water bath, squeeze the bellow and point the outlet to a bucket or sink
Specifications
Specifications
| For Use With (Equipment) | NE4-8 D/CT, NE1B-28, NE1-2.5 E, NE4-28 D, NE4-22 T Stirred Digital Clear Water Baths |
| Temperature Range | Below 45°C |
| Type | Draining Syphon |