Learn More
Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped with glass plug
Product Code. p-7116293
Click to view available options
Capacity (Metric):
50 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Product Code. 11876074
Brand: Lenz Laborglasinstrumente™ 04452802
Only null left
Add to basket
Add to basket
Made from borosilicate glass. Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped with glass plug is used in the laboratory to transfer fluids. The funnel is fitted with a stopcock which allows the flow to be controlled.
  • Conical shape with NS socket and NS cone
  • With solid glass plug, with screwthread retaining device
  • With inscription label, without scale
  • Borosilicate glass
Specifications
Capacity (Metric) 50 mL
Material Borosilicate Glass
Pore Size 2.5 mm
Length (Metric) Overall 100 mm